Merck
Potent inhibitor scaffold against Trypanosoma cruzi trans-sialidase.
Bioorganic & medicinal chemistry (2010-01-26)
Shingo Arioka, Masahiro Sakagami, Rie Uematsu, Hiroto Yamaguchi, Hiroko Togame, Hiroshi Takemoto, Hiroshi Hinou, Shin-Ichiro Nishimura
ABSTRACT
The protozoan Trypanosoma cruzi, the causative agent of Chagas' disease, can infect the heart, causing cardiac arrest frequently followed by death. To treat this disease, a potential molecular drug target is T. cruzi trans-sialidase (TcTS). However, inhibitors found to date are not strong enough to serve as a lead scaffold; most inhibitors reported thus far are derivatives of the substrate sialic acid or a transition state analogue known as 2,3-dehydro-3-deoxy-N-acetylneuraminic acid (DANA) with an IC(50) value of more than hundreds of micromolar. Since natural products are highly stereodiversified and often provide highly specific biological activity, we screened a natural product library for inhibitors of TcTS and identified promising flavonoid and anthraquinone derivatives. A structure-activity relationship (SAR) analysis of the flavonoids revealed that apigenin had the minimal and sufficient structure for inhibition. Intriguingly, the compound has been reported to possess trypanocidal activity. An SAR analysis of anthraquinones showed that 6-chloro-9,10-dihydro-4,5,7-trihydroxy-9,10-dioxo-2-anthracenecarboxylic acid had the strongest inhibitory activity ever found against TcTS. Moreover, its inhibitory activity appeared to be specific to TcTS. These compounds may serve as potent lead chemotherapeutic scaffolds against Chagas' disease.
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Rhein
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
(−)-Catechin, ≥97% (HPLC), from green tea
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Emodin, from Frangula bark, ≥90% (HPLC)
Sigma-Aldrich
Chrysin, ≥96.5%