Merck
  • Clinical Routine Application of the Second-generation Neuroendocrine Markers ISL1, INSM1, and Secretagogin in Neuroendocrine Neoplasia: Staining Outcomes and Potential Clues for Determining Tumor Origin.
Clinical Routine Application of the Second-generation Neuroendocrine Markers ISL1, INSM1, and Secretagogin in Neuroendocrine Neoplasia: Staining Outcomes and Potential Clues for Determining Tumor Origin.
Endocrine pathology (2020-08-20)
Carl Christofer Juhlin, Jan Zedenius, Anders Höög
ABSTRACT
Neuroendocrine neoplasms (NENs) have traditionally been identified via expression of proteins associated to the regulation of secretory vesicles and granules. We report the clinical usage of the "second-generation" proteins ISL LIM homeobox 1 (ISL1), INSM transcriptional repressor 1 (INSM1), and secretagogin (SECG) as immunohistochemical markers of neuroendocrine differentiation since their introduction in clinical routine and compare the results with the established proteins chromogranin A (CGA) and synaptophysin (SYP). In total, 161 tumors, including 139 NENs and 22 "non-NENs" (unrelated tumors with an initial suspicion of NEN), were informatively stained for ISL1, and subsets were also interrogated for INSM1 and/or SECG. Diffuse or focal positive immunoreactivity was noted for ISL1 in 91/139 NENs (65%) and in 6/22 (27%) non-NENs, for INSM1 in 76/85 NENs (89%) and in 2/5 (40%) non-NENs, and for SECG in 49 out of 64 NENs (77%) and in 0/5 non-NENs (0%). Generally, ISL1, INSM1, and SECG exhibited sensitivities in line with or slightly below that of CGA and SYP-largely attributable to tissue-specific patterns regarding tumoral origin. Moreover, for pancreatic and small intestinal NENs, the two largest subgroups, ISL1 staining results were consistent irrespectively of tumor source and WHO grade. We verify previously suggested immunohistochemical schemes of neuroendocrine markers of first- and second-generations to facilitate the diagnostic process for NENs and confirm that the second-generation neuroendocrine markers display tissue-specific patterns. We therefore recommend their implementation in tertiary endocrine pathology centers, not least to aid in the identification of primary tumors when analyzing metastases.