Merck
  • Metabolic degradation of lentinan in liver mediated by CYP450 enzymes and epoxide hydrolase.
Metabolic degradation of lentinan in liver mediated by CYP450 enzymes and epoxide hydrolase.
Carbohydrate polymers (2020-12-07)
Ziming Zheng, Yu Zhang, Yuxuan Liu, Jinglin Wang, Zheng Cui, Xianglin Pan, Yan Liu, Wenqi Tang, Kaiping Wang
ABSTRACT
Lentinan (LNT), a typical triple helix β-glucan, has been widely used as drug and biomaterial. However, its pharmacokinetics in vivo is rarely reported, which severely limits its further development and application. The aim of this study is to establish a sensitive method for detecting LNT in biosamples and to evaluate the plasma level, tissue distribution and metabolic degradation of LNT in rats. 5-([4,6-Dichlorotriazin-2-yl] amino) fluorescein (DTAF) was labelled to LNT. After purification and identification, FLNT was intravenously administered to rats at dose of 32 mg/kg. LNT was predominantly incorporated into the liver and liver microsomes were used to study the degradation mechanism of LNT in the liver. The results showed that two cytochrome P450 (CYP450) enzymes subtypes (CYP2D6 and CYP2C9), as well as epoxide hydrolase, were involved in the metabolic degradation of LNT. These findings provide a pharmacokinetic reference for further study and application of LNT and other β-glucans.
Sigma-Aldrich
Valpromide, ≥97% (NMR)