Merck
  • Structural insight into human CK2alpha in complex with the potent inhibitor ellagic acid.
Structural insight into human CK2alpha in complex with the potent inhibitor ellagic acid.
Bioorganic & medicinal chemistry letters (2009-05-06)
Yusuke Sekiguchi, Tetsuko Nakaniwa, Takayoshi Kinoshita, Isao Nakanishi, Kazuo Kitaura, Akira Hirasawa, Gozoh Tsujimoto, Toshiji Tada
ABSTRACT
We determined the 2.35-A crystal structure of a human CK2 catalytic subunit (referred to as CK2alpha complexed with the ATP-competitive, potent CK2 inhibitor ellagic acid. The inhibitor binds to CK2alpha with a novel binding mode, including water-mediated hydrogen bonds. This structural information may support discovery of potent CK2 inhibitors.
Sigma-Aldrich
Emodin, from Frangula bark, ≥90% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)