Merck
  • Isocombretastatins a versus combretastatins a: the forgotten isoCA-4 isomer as a highly promising cytotoxic and antitubulin agent.
Isocombretastatins a versus combretastatins a: the forgotten isoCA-4 isomer as a highly promising cytotoxic and antitubulin agent.
Journal of medicinal chemistry (2009-06-18)
Samir Messaoudi, Bret Tréguier, Abdallah Hamze, Olivier Provot, Jean-François Peyrat, Jordi Rodrigo De Losada, Jian-Miao Liu, Jérôme Bignon, Joanna Wdzieczak-Bakala, Sylviane Thoret, Joëlle Dubois, Jean-Daniel Brion, Mouâd Alami
ABSTRACT
Herein is reported a convergent synthesis of isocombretastatins A, a novel class of potent antitubulin agents. These compounds having a 1,1-diarylethylene scaffold constitute the simplest isomers of natural Z-combretastatins A that are easy to synthesize without need to control the Z-olefin geometry. The discovery of isoCA-4 with biological activities comparable to that of CA-4 represents a major progress in this field.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)