Merck
  • Neurosteroid analogues. 12. Potent enhancement of GABA-mediated chloride currents at GABAA receptors by ent-androgens.
Neurosteroid analogues. 12. Potent enhancement of GABA-mediated chloride currents at GABAA receptors by ent-androgens.
European journal of medicinal chemistry (2007-04-17)
Bryson W Katona, Kathiresan Krishnan, Zu Yun Cai, Brad D Manion, Ann Benz, Amanda Taylor, Alex S Evers, Charles F Zorumski, Steven Mennerick, Douglas F Covey
ABSTRACT
Allopregnanolone (1) and pregnanolone (2), steroids containing a 17beta-acetyl group, are potent enhancers of GABA (gamma-aminobutyric acid) action at GABAA receptors. Their effects are enantioselective with the non-naturally occurring enantiomers (ent-1 and ent-2) being less potent. Androsterone (3) and etiocholanolone (4), steroids with a C-17 carbonyl group, are weak enhancers of GABA action at GABAA receptors. Unexpectedly, their enantiomers (ent-3 and ent-4) have been found to have enhanced, not diminished, activity at GABAA receptors. Furthermore, the C-17 spiro-epoxide analogues (ent-5 and ent-6) of ent-3 and ent-4, respectively, have activities comparable to those of steroids 1 and 2. The results indicate that some ent-steroids are potent modulators of GABAA receptors and might have clinical potential as GABAergic drugs of the future.
Product Description
Supelco
Androsterone, VETRANAL®, analytical standard
Sigma-Aldrich
Etiocholan-3α-ol-17-one
Sigma-Aldrich
5α-Pregnan-3α-ol-20-one, solid