Merck
Immunofluorescence staining.
Current protocols in cell biology (2008-01-30)
J G Donaldson
ABSTRACT
This unit provides a protocol for indirect immunofluorescence, which is a method that provides information about the locations of specific molecules and the structure of the cell. Antibody molecules for a specific target molecule are exposed to the cell or tissue being investigated. The binding of these molecules is detected by incubating the sample with a secondary antibody specific for immunoglobulin molecules and conjugated to fluorophore. This provides both a visible signal and amplification of the signal and the results are observed with a fluorescent microscope.