Merck
  • Development, validation, and application of a quantitative volumetric absorptive microsampling-based method in finger prick blood by means of LC-HRMS/MS applicable for adherence monitoring of antipsychotics.
Development, validation, and application of a quantitative volumetric absorptive microsampling-based method in finger prick blood by means of LC-HRMS/MS applicable for adherence monitoring of antipsychotics.
Analytical and bioanalytical chemistry (2021-02-01)
Cathy M Jacobs, Lea Wagmann, Markus R Meyer
ABSTRACT
Volumetric absorptive microsampling (VAMS), an emerging microsampling technique, is expected to overcome some disadvantages of dried blood spots such as volume inaccuracy and influence of hematocrit (HT). This study aimed to develop and evaluate a VAMS-based strategy for quantification of 13 frequently prescribed antipsychotics in finger prick blood within the scope of adherence monitoring to complement already-established qualitative urine analysis. The final workflow consisted of VAMS tip hydration and subsequent precipitation. Samples were analyzed by using reversed-phase ultra-high-performance liquid chromatography and Orbitrap mass spectrometry operated in parallel reaction monitoring mode. The analytical procedure was successfully validated based on international recommendations at three different HT values (20%, 40%, 60%) for most of the analytes. Selectivity and within/between-run accuracy and precision were in accordance with the recommendations in most cases. Internal standard-normalized matrix factor met recommended criteria for all analytes at HT 40%. For the HT values of 20% and 60%, only four substances did not meet the criteria. Dilution integrity was given for all substances, except for olanzapine, allowing a quantification over the whole therapeutic range of selected antipsychotics. Long-term stability in VAMS tips was tested and revealed degradation of five antipsychotic drugs after 1 week of storage at 24 °C. A proof of concept of the applicability of the method was obtained by quantification of a selection of the 13 antipsychotic drugs in VAMS tips and matched plasma samples. Results were coherent between matrices. Thus, VAMS was shown to be a promising alternative for adherence monitoring of at least the investigated antipsychotics.
Sigma-Aldrich
Cyamemazine, ≥98% (HPLC)