Merck
Mechanisms, biology and inhibitors of deubiquitinating enzymes.
Nature chemical biology (2007-10-20)
Kerry Routenberg Love, André Catic, Christian Schlieker, Hidde L Ploegh
ABSTRACT
The addition of ubiquitin (Ub) and ubiquitin-like (Ubl) modifiers to proteins serves to modulate function and is a key step in protein degradation, epigenetic modification and intracellular localization. Deubiquitinating enzymes and Ubl-specific proteases, the proteins responsible for the removal of Ub and Ubls, act as an additional level of control over the ubiquitin-proteasome system. Their conservation and widespread occurrence in eukaryotes, prokaryotes and viruses shows that these proteases constitute an essential class of enzymes. Here, we discuss how chemical tools, including activity-based probes and suicide inhibitors, have enabled (i) discovery of deubiquitinating enzymes, (ii) their functional profiling, crystallographic characterization and mechanistic classification and (iii) development of molecules for therapeutic purposes.
Product Description
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications