Merck
Anti-mycobacterial activity of a bis-sulfonamide.
Bioorganic & medicinal chemistry letters (2007-01-30)
Brendan L Wilkinson, Laurent F Bornaghi, Anthony D Wright, Todd A Houston, Sally-Ann Poulsen
ABSTRACT
A bis-arylsulfonamide, 7, has been identified that exhibits growth inhibition of Mycobacterium smegmatis at less than 25 microg/mL, but has no such activity against Escherichia coli or Staphylococcus aureus. A closely related bis-arylsulfonamide (8) was much less active, but was the only other compound among 54 arylsulfonamides tested with detectable growth inhibition of M. smegmatis.
Sigma-Aldrich
4-Sulfamoylbenzoic acid, 97%
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)