Merck
  • The Oxygen Reduction Reaction in Ca2+ -Containing DMSO: Reaction Mechanism, Electrode Surface Characterization, and Redox Mediation*.
The Oxygen Reduction Reaction in Ca2+ -Containing DMSO: Reaction Mechanism, Electrode Surface Characterization, and Redox Mediation*.
ChemSusChem (2020-09-01)
Pawel Peter Bawol, Philip Heinrich Reinsberg, Andreas Koellisch-Mirbach, Christoph Johannes Bondue, Helmut Baltruschat
ABSTRACT
In this study the fundamental understanding of the underlying reactions of a possible Ca-O2 battery using a DMSO-based electrolyte was strengthened. Employing the rotating ring disc electrode, a transition from a mixed process of O2 - and O2 2- formation to an exclusive O2 - formation at gold electrodes is observed. It is shown that in this system Ca-superoxide and Ca-peroxide are formed as soluble species. However, there is a strongly adsorbed layer of products of the oxygen reduction reaction (ORR) s on the electrode surface, which is blocking the electrode. Surprisingly the blockade is only a partial blockade for the formation of peroxide while the formation of superoxide is maintained. During an anodic sweep, the ORR product layer is stripped from the electrode surface. With X-ray photoelectron spectroscopy (XPS) the deposited ORR products were shown to be Ca(O2 )2 , CaO2 , and CaO as well as side-reaction products such as CO3 2- and other oxygen-containing carbon species. It is shown that the strongly attached layer on the electrocatalyst, that was partially blocking the electrode, could be adsorbed CaO. The disproportionation reaction of O2 - in presence of Ca2+ was demonstrated via mass spectrometry. Finally, the ORR mediated by 2,5-di-tert-1,4-benzoquinone (DBBQ) was investigated by differential electrochemical mass spectrometry (DEMS) and XPS. Similar products as without DBBQ are deposited on the electrode surface. The analysis of the DEMS experiments shows that DBBQ- reduces O2 to O2 - and O2 2- , whereas in the presence of DBBQ2- O2 2- is formed. The mechanism of the ORR with and without DBBQ is discussed.
Sigma-Aldrich
Calcium perchlorate tetrahydrate, 99%