Merck
  • Preventive effect of exercise training on diabetic kidney disease in ovariectomized rats with type 1 diabetes.
Preventive effect of exercise training on diabetic kidney disease in ovariectomized rats with type 1 diabetes.
Experimental biology and medicine (Maywood, N.J.) (2019-05-02)
Cláudia Silva Souza, Bianca Silva de Sousa Oliveira, Geovanildo Nascimento Viana, Thiago Macêdo Lopes Correia, Ana Carolina de Bragança, Daniele Canale, Márcio Vasconcelos Oliveira, Amélia Cristina Mendes de Magalhães, Rildo Aparecido Volpini, Liliany Souza de Brito Amaral, Telma de Jesus Soares
ABSTRACT
To date, no studies have been found evaluating the effects of physical exercise on renal function and structure changes in ovariectomized rats with type 1 diabetes. Therefore, this work emerges with an important tool for strengthening and expanding innovative research on exercise with potential for the prevention of renal diseases in ovariectomized diabetic rats, and future development of studies that seek to increase scientific knowledge about the beneficial effects of physical exercise on renal diseases in humans.
Sigma-Aldrich
Anti-Fibronectin Antibody, Chemicon®, from rabbit