Merck
  • G-Protein types involved in calcium channel inhibition at a presynaptic nerve terminal.
G-Protein types involved in calcium channel inhibition at a presynaptic nerve terminal.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2000-10-12)
R R Mirotznik, X Zheng, E F Stanley
ABSTRACT
The inhibition of presynaptic calcium channels via G-protein-dependent second messenger pathways is a key mechanism of transmitter release modulation. We used the calyx-type nerve terminal of the chick ciliary ganglion to examine which G-proteins are involved in the voltage-sensitive inhibition of presynaptic N-type calcium channels. Adenosine caused a prominent inhibition of the calcium current that was totally blocked by pretreatment with pertussis toxin (PTX), consistent with an exclusive involvement of G(o)/G(i) in the G-protein pathway. Immunocytochemistry was used to localize these G-protein types to the nerve terminal and its transmitter release face. We used two approaches to test for modulation by other G-protein types. First, we treated the terminals with ligands for a variety of G-protein-linked neurotransmitter receptor types that have been associated with different G-protein families. Although small inhibitory effects were observed, these could all be eliminated by PTX, indicating that in this terminal the G(i) family is the sole transmitter-induced G-protein inhibitory pathway. Second, we examined the kinetics of calcium channel inhibition by uncaging the nonselective and irreversible G-protein activator GTPgammaS, bypassing the receptors. A large fraction of the rapid GTPgammaS-induced inhibition persisted, consistent with a G(o)/G(i)-independent pathway. Immunocytochemistry identified G(q), G(11), G(12), and G(13) as potential PTX-insensitive second messengers at this terminal. Thus, our results suggest that whereas neurotransmitter-mediated calcium channel inhibition is mainly, and possibly exclusively, via G(o)/G(i), other rapid PTX-insensitive G-protein pathways exist that may involve novel, and perhaps transmitter-independent, activating mechanisms.
Sigma-Aldrich
Anti-Giα-3-Subunit, C-Terminal (345-354) Rabbit pAb, liquid, Calbiochem®