Merck
  • CT10: a new cancer-testis (CT) antigen homologous to CT7 and the MAGE family, identified by representational-difference analysis.
CT10: a new cancer-testis (CT) antigen homologous to CT7 and the MAGE family, identified by representational-difference analysis.
International journal of cancer (2000-03-04)
A O Güre, E Stockert, K C Arden, A D Boyer, C S Viars, M J Scanlan, L J Old, Y T Chen
ABSTRACT
Assays relying on humoral or T-cell-based recognition of tumor antigens to identify potential targets for immunotherapy have led to the discovery of a significant number of immunogenic gene products, including cancer-testis (CT) antigens predominantly expressed in cancer cells and male germ cells. The search for cancer-specific antigens has been extended via the technique of representational-difference analysis and SK-MEL-37, a melanoma cell line expressing a broad range of CT antigens. Using this approach, we have isolated CT antigen genes, genes over-expressed in cancer, e. g., PRAME and KOC, and genes encoding neuro-ectodermal markers. The identified CT antigen genes include the previously defined MAGE-A6, MAGE-A4a, MAGE-A10, CT7/MAGE-C1, as well as a novel gene designated CT10, which shows strong homology to CT7/MAGE-C1 both at cDNA and at genomic levels. Chromosome mapping localized CT10 to Xq27, in close proximity to CT7/MAGE-C1 and MAGE-A genes. CT10 mRNA is expressed in testis and in 20 to 30% of various human cancers. A serological survey identified 2 melanoma patients with anti-CT10 antibody, demonstrating the immunogenicity of CT10 in humans.