Merck
  • Design and synthesis of substrate and intermediate analogue inhibitors of S-ribosylhomocysteinase.
Design and synthesis of substrate and intermediate analogue inhibitors of S-ribosylhomocysteinase.
Journal of medicinal chemistry (2006-05-12)
Gang Shen, Rakhi Rajan, Jinge Zhu, Charles E Bell, Dehua Pei
ABSTRACT
S-Ribosylhomocysteinase (LuxS) catalyzes the cleavage of the thioether linkage in S-ribosylhomocysteine (SRH) to produce homocysteine and 4,5-dihydroxy-2,3-pentanedione, the precursor of autoinducer 2. Inhibitors of LuxS should interfere with bacterial interspecies communication and potentially provide a novel class of antibacterial agents. LuxS utilizes a divalent metal ion as a Lewis acid during catalysis. In this work, a series of structural analogues of the substrate SRH and a 2-ketone intermediate were designed and synthesized. Kinetic studies indicate that the compounds act as reversible, competitive inhibitors against LuxS, with the most potent inhibitors having K(I) values in the submicromolar range. These represent the most potent LuxS inhibitors that have been reported to date. Cocrystal structures of LuxS bound with two of the inhibitors largely confirmed the design principles, i.e., the importance of both the homocysteine and ribose moieties in high-affinity binding to the LuxS active site.
Product Description
SAFC
L-Methionine
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)