Merck
  • Calcium-Mediated Liposome Fusion to Engineer Giant Lipid Vesicles with Cytosolic Proteins and Reconstituted Mammalian Proteins.
Calcium-Mediated Liposome Fusion to Engineer Giant Lipid Vesicles with Cytosolic Proteins and Reconstituted Mammalian Proteins.
Advanced biosystems (2020-10-22)
Yannick R F Schmid, Leo Scheller, Sebastian Buchmann, Petra S Dittrich
ABSTRACT
Giant unilamellar lipid vesicles (GUVs) are widely used as model membrane systems and provide an excellent basis to construct artificial cells. To construct more sophisticated artificial cells, proteins-in particular membrane proteins-need to be incorporated in GUVs. However, current methods for protein reconstitution have limited throughput or are not generally applicable for all proteins because they depend on detergent solubilization. This limitation is addressed here by introducing calcium-mediated membrane fusion to transfer proteins between negatively charged GUVs and cell-derived plasma membrane vesicles (CDVs), derived from HEK293T cells overexpressing a membrane receptor protein. Fusion conditions are optimized using large unilamellar vesicles and GUVs containing phosphatidylserines and fusogenic lipids. The approach is then applied to induce lipid mixing and subsequent transfer of the overexpressed membrane receptor from CDVs into GUVs. The membrane receptor is detected by immunofluorescence on GUVs that underwent lipid mixing with CDVs. Those GUVs also exhibit esterase activity because cytosolic esterases entrapped in the CDVs are transferred during membrane fusion. Thus, content mixing is demonstrated. Using CDVs circumvents the need to purify or solubilize proteins. Moreover, calcium-mediated fusion allows transfer of lipids, water-soluble and membrane bound proteins in one step, resulting in a semi-synthetic cell.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
Mineral oil, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.