- Type-IInterferon-Inducible SERTAD3 Inhibits Influenza A Virus Replication by Blocking the Assembly of Viral RNA Polymerase Complex.
Type-IInterferon-Inducible SERTAD3 Inhibits Influenza A Virus Replication by Blocking the Assembly of Viral RNA Polymerase Complex.
Influenza A virus (IAV) infection stimulates a type I interferon (IFN-I) response in host cells that exerts antiviral effects by inducing the expression of hundreds of IFN-stimulated genes (ISGs). However, most ISGs are poorly studied for their roles in the infection of IAV. Herein, we demonstrate that SERTA domain containing 3 (SERTAD3) has a significant inhibitory effect on IAV replication in vitro. More importantly, Sertad3-/- mice develop more severe symptoms upon IAV infection. Mechanistically, we find SERTAD3 reduces IAV replication through interacting with viral polymerase basic protein 2 (PB2), polymerase basic protein 1 (PB1), and polymerase acidic protein (PA) to disrupt the formation of the RNA-dependent RNA polymerase (RdRp) complex. We further identify an 8-amino-acid peptide of SERTAD3 as a minimum interacting motif that can disrupt RdRp complex formation and inhibit IAV replication. Thus, our studies not only identify SERTAD3 as an antiviral ISG, but also provide the mechanism of potential application of SERTAD3-derived peptide in suppressing influenza replication.