Merck
Anti-HIV-Active Nucleoside Triphosphate Prodrugs.
Journal of medicinal chemistry (2020-05-19)
Xiao Jia, Dominique Schols, Chris Meier
ABSTRACT
We disclose a study on nucleoside triphosphate (NTP) analogues in which the γ-phosphate is covalently modified by two different biodegradable masking units and d4T as nucleoside analogue that enable the delivery of d4TTP with high selectivity in phosphate buffer (pH 7.3) and by enzyme-triggered reactions in human CD4+ T-lymphocyte CEM cell extracts. This allows the bypass of all steps normally needed in the intracellular phosphorylation. These TriPPPro-nucleotides comprising an acyloxybenzyl (AB; ester) or an alkoxycarbonyloxybenzyl (ACB; carbonate) in combination with an ACB moiety are described as NTP delivery systems. The introduction of these two different groups led to the selective formation of γ-(ACB)-d4TTPs by chemical hydrolysis and in particular by cell extract enzymes. γ-(AB)-d4TTPs are faster cleaved than γ-(ACB)-d4TTPs. In antiviral assays, the compounds are highly active against HIV-1 and HIV-2 in wild-type CEM/O cells and more importantly in thymidine kinase-deficient CD4+ T-cells (CEM/TK-).
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%