Merck
  • Small molecule H89 renders the phosphorylation of S6K1 and AKT resistant to mTOR inhibitors.
Small molecule H89 renders the phosphorylation of S6K1 and AKT resistant to mTOR inhibitors.
The Biochemical journal (2020-04-30)
Chase H Melick, Jenna L Jewell
ABSTRACT
The mammalian target of rapamycin (mTOR) is an evolutionarily conserved Ser/Thr kinase that comprises two complexes, termed mTOR complex 1 (mTORC1) and mTOR complex 2 (mTORC2). mTORC1 phosphorylates S6K1 at Thr 389, whereas mTORC2 phosphorylates AKT at Ser 473 to promote cell growth. As the mTOR name implies it is the target of natural product called rapamycin, a clinically approved drug used to treat human disease. Short-term rapamycin treatment inhibits the kinase activity of mTORC1 but not mTORC2. However, the ATP-competitive catalytic mTOR inhibitor Torin1 was identified to inhibit the kinase activity of both mTORC1 and mTORC2. Here, we report that H89 (N-(2-(4-bromocinnamylamino) ethyl)-5-isoquinolinesulfonamide), a well-characterized ATP-mimetic kinase inhibitor, renders the phosphorylation of S6K1 and AKT resistant to mTOR inhibitors across multiple cell lines. Moreover, H89 prevented the dephosphorylation of AKT and S6K1 under nutrient depleted conditions. PKA and other known H89-targeted kinases do not alter the phosphorylation status of S6K1 and AKT. Pharmacological inhibition of some phosphatases also enhanced S6K1 and AKT phosphorylation. These findings suggest a new target for H89 by which it sustains the phosphorylation status of S6K1 and AKT, resulting in mTOR signaling.
Sigma-Aldrich
c-Myc Peptide, lyophilized powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Insulin human, ≥95% (HPLC), semisynthetic, powder, non-sterile
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
MISSION® esiRNA, targeting human PPP1CA