Merck
  • Engineered Polymersomes for the Treatment of Fish Odor Syndrome: A First Randomized Double Blind Olfactory Study.
Engineered Polymersomes for the Treatment of Fish Odor Syndrome: A First Randomized Double Blind Olfactory Study.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2020-04-25)
Aaron C Schmidt, Erik R Hebels, Charlotte Weitzel, Anna Kletzmayr, Yinyin Bao, Christian Steuer, Jean-Christophe Leroux
ABSTRACT
Trimethylamine (TMA) is a metabolite overtly present in patients suffering from trimethylaminuria (TMAU), a rare genetic disorder characterized by a strong "fishy" body odor. To date, no approved pharmacological treatment to sequester excess TMA on the skin of patients exists. Here, transmembrane pH gradient poly(isoprene)-block-poly(ethylene glycol) (PI-b-PEG) polymersomes are investigated for the topical removal of TMA. PI-b-PEG amphiphiles of varying chain length are synthesized and evaluated for their ability to form vesicular structures in aqueous media. The optimization of the PI/PEG ratio of transmembrane pH gradient polymersomes allows for the rapid and efficient capture of TMA both in solution and after incorporation into a topical hydrogel matrix at the pH of the skin. A subsequent double blind olfactory study reveals a significant decrease in perceived odor intensity after application of the polymersome-based formulation on artificial skin substrates that has been incubated in TMA-containing medium. This simple and novel approach has the potential to ease the burden of people suffering from TMAU.
Sigma-Aldrich
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%