Merck
  • Molecular Analysis of Lipid-Reactive Vδ1 γδ T Cells Identified by CD1c Tetramers.
Molecular Analysis of Lipid-Reactive Vδ1 γδ T Cells Identified by CD1c Tetramers.
Journal of immunology (Baltimore, Md. : 1950) (2016-01-13)
Sobhan Roy, Dalam Ly, Caitlin D Castro, Nan-Sheng Li, Andrew J Hawk, John D Altman, Stephen C Meredith, Joseph A Piccirilli, D Branch Moody, Erin J Adams
ABSTRACT
CD1c is abundantly expressed on human dendritic cells (DC) and B cells, where it binds and displays lipid Ags to T cells. In this study, we report that CD1c tetramers carrying Mycobacterium tuberculosis phosphomycoketide bind γδ TCRs. An unbiased method of ligand-based TCR selection detects interactions only with Vδ1(+) TCRs, and mutational analyses demonstrate a role of the Vδ1 domain during recognition. These results strengthen evidence for a role of CD1c in the γδ T cell response, providing biophysical evidence for CD1c-γδ TCR interactions and a named foreign Ag. Surprisingly, TCRs also bind CD1c complexes formed with diverse lipids such as lysophosphatidylcholine, sulfatide, or mannosyl-phosophomycoketide, but not lipopeptide ligands. Dissection of TCR interactions with CD1c carrying foreign Ags, permissive ligands, and nonpermissive lipid ligands clarifies the molecular basis of the frequently observed but poorly understood phenomenon of mixed self- and foreign Ag reactivity in the CD1 system.
Product Description
Avanti
18:1 Lyso PC, 1-oleoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform