Merck
  • Further antinociceptive effects of myricitrin in chemical models of overt nociception in mice.
Further antinociceptive effects of myricitrin in chemical models of overt nociception in mice.
Neuroscience letters (2011-02-15)
Marina Machado Córdova, Maria Fernanda de Paula Werner, Morgana Duarte da Silva, Ana Paula Ruani, Moacir Geraldo Pizzolatti, Adair R S Santos
ABSTRACT
The present work explored the antinociceptive effects of the flavonoid myricitrin in models of overt nociception triggered by intraplantar injection of chemical algogens into the hind paw of mice. The nociception induced by bradykinin (3 nmol/paw i.pl.) was abolished by prior treatment with myricitrin (10-100mg/kg, i.p.) with ID(50) of 12.4 (8.5-18.1)mg/kg. In sharp contrast, myricitrin failed to affect the nociception elicited by prostaglandin E(2) (3 nmol/paw i.pl.). Cinnamaldehyde (10 nmol/paw i.pl.)-induced nociception was reduced by myricitrin (100mg/kg, i.p.) and camphor (7.6 mg/kg,s.c.) in 43±10% and 57±8%, respectively. Myricitrin (30-100mg/kg, i.p.) and amiloride (100mg/kg, i.p.) inhibited nociceptive responses induced by acidified saline (pH 5/paw i.pl.), with ID(50) of 22.0 (16.1-30.0)mg/kg and inhibition of 71±6% and 64±5%, respectively. Moreover, myricitrin (10-30 mg/kg, i.p.) and ruthenium red (3mg/kg, i.p.) significantly reduced the nociception induced by menthol (1.2 μmol/paw i.pl.) with the mean ID(50) of 2.4 (1.5-3.7)mg/kg and inhibition of 95±3% and 51±7%, respectively. In addition, myricitrin administration (30 and 100mg/kg, i.p.) markedly reduced menthol-induced mechanical allodynia. However, myricitrin (100mg/kg, i.p.) prevented (only in time of 60 min) cold allodynia induced by menthol. Collectively, the present results extend prior data and show that myricitrin promotes potent antinociception, an action that is likely mediated by an inhibition of the activation of nociceptors by bradykinin and TRPs agonist (i.e. cinnamaldehyde, acidified saline and menthol), probably via inhibition of PKC pathways. Thus, myricitrin could constitute an attractive molecule of interest for the development of new analgesic drugs.
Sigma-Aldrich
Myricitrin, ≥99.0% (HPLC)
Supelco
Myricitrin, analytical standard