Merck
  • Anti-fibrotic effects of nintedanib in lung fibroblasts derived from patients with idiopathic pulmonary fibrosis.
Anti-fibrotic effects of nintedanib in lung fibroblasts derived from patients with idiopathic pulmonary fibrosis.
Respiratory research (2014-12-17)
Katrin E Hostettler, Jun Zhong, Eleni Papakonstantinou, George Karakiulakis, Michael Tamm, Petra Seidel, Qingzhu Sun, Jyotshna Mandal, Didier Lardinois, Christopher Lambers, Michael Roth
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a progressive lung disease with poor prognosis. The kinase inhibitor nintedanib specific for vascular endothelial growth factor receptor (VEGFR), platelet-derived growth factor receptor (PDGFR) and fibroblast growth factor receptor (FGFR) significantly reduced the rate of decline of forced vital capacity versus placebo. To determine the in vitro effect of nintedanib on primary human lung fibroblasts. Fibroblasts were isolated from lungs of IPF patients and from non-fibrotic controls. We assessed the effect of VEGF, PDGF-BB and basic FGF (bFGF) ± nintedanib on: (i) expression/activation of VEGFR, PDGFR, and FGFR, (ii) cell proliferation, secretion of (iii) matrix metalloproteinases (MMP), (iv) tissue inhibitor of metalloproteinase (TIMP), and (v) collagen. IPF fibroblasts expressed higher levels of PDGFR and FGFR than controls. PDGF-BB, bFGF, and VEGF caused a pro-proliferative effect which was prevented by nintedanib. Nintedanib enhanced the expression of pro-MMP-2, and inhibited the expression of TIMP-2. Transforming growth factor-beta-induced secretion of collagens was inhibited by nintedanib. Our data demonstrate a significant anti-fibrotic effect of nintedanib in IPF fibroblasts. This effect consists of the drug's anti-proliferative capacity, and on its effect on the extracellular matrix, the degradation of which seems to be enhanced.
Sigma-Aldrich
Nintedanib ethanesulphonate, ≥98% (HPLC)
Sigma-Aldrich
Nintedanib, ≥98% (HPLC)