Merck
  • α-Ketoglutarate-Activated NF-κB Signaling Promotes Compensatory Glucose Uptake and Brain Tumor Development.
α-Ketoglutarate-Activated NF-κB Signaling Promotes Compensatory Glucose Uptake and Brain Tumor Development.
Molecular cell (2019-08-27)
Xiongjun Wang, Ruilong Liu, Xiujuan Qu, Hua Yu, Huiying Chu, Yajuan Zhang, Wencheng Zhu, Xueyuan Wu, Hong Gao, Bangbao Tao, Wenfeng Li, Ji Liang, Guohui Li, Weiwei Yang
ABSTRACT
The rapid proliferation of cancer cells and dysregulated vasculature within the tumor leads to limited nutrient accessibility. Cancer cells often rewire their metabolic pathways for adaption to nutrient stress, and the underlying mechanism remains largely unknown. Glutamate dehydrogenase 1 (GDH1) is a key enzyme in glutaminolysis that converts glutamate to α-ketoglutarate (α-KG). Here, we show that, under low glucose, GDH1 is phosphorylated at serine (S) 384 and interacts with RelA and IKKβ. GDH1-produced α-KG directly binds to and activates IKKβ and nuclear factor κB (NF-κB) signaling, which promotes glucose uptake and tumor cell survival by upregulating GLUT1, thereby accelerating gliomagenesis. In addition, GDH1 S384 phosphorylation correlates with the malignancy and prognosis of human glioblastoma. Our finding reveals a unique role of α-KG to directly regulate signal pathway, uncovers a distinct mechanism of metabolite-mediated NF-κB activation, and also establishes the critical role of α-KG-activated NF-κB in brain tumor development.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Phosphoserine Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
L-Glutamic acid monosodium salt monohydrate, ≥98.0% (NT)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Octyl-(R)-2HG, ≥98% (HPLC)
Sigma-Aldrich
Octyl-α-KG, ≥95% (HPLC)
Sigma-Aldrich
TNF- α Protein, Human Recombinant Animal Free, Tumor necrosis factor alpha (TNF-α), also known as cachectin & TNFSF1A, is the prototypic ligand of the TNF superfamily. Recombinant animal free human TNF-α is manufactured using all non-animal reagents.