Merck
  • Adenosine enhances progenitor cell recruitment and nerve growth via its A2B receptor during adult fin regeneration.
Adenosine enhances progenitor cell recruitment and nerve growth via its A2B receptor during adult fin regeneration.
Purinergic signalling (2014-08-03)
Christine Rampon, Carole Gauron, Francesca Meda, Michel Volovitch, Sophie Vriz
ABSTRACT
A major issue in regenerative medicine is the control of progenitor cell mobilisation. Apoptosis has been reported as playing a role in cell plasticity, and it has been recently shown that apoptosis is necessary for organ and appendage regeneration. In this context, we explore its possible mode of action in progenitor cell recruitment during adult regeneration in zebrafish. Here, we show that apoptosis inhibition impairs blastema formation and nerve growth, both of which can be restored by exogenous adenosine acting through its A2B receptor. Moreover, adenosine increases the number of progenitor cells. Purinergic signalling is therefore an early and essential event in the pathway from lesion to blastema formation and provides new targets for manipulating cell plasticity in the adult.
Sigma-Aldrich
CGS-21680 hydrochloride hydrate, solid
Sigma-Aldrich
MRS 1754 hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Apoptosis Inhibitor II, NS3694, The Apoptosis Inhibitor II, NS3694, also referenced under CAS 426834-38-0, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Apoptosis Inhibitor, The Apoptosis Inhibitor, also referenced under CAS 54135-60-3, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.