Merck
  • One-pot synthesis of chlorhexidine-templated biodegradable mesoporous organosilica nanoantiseptics.
One-pot synthesis of chlorhexidine-templated biodegradable mesoporous organosilica nanoantiseptics.
Colloids and surfaces. B, Biointerfaces (2019-12-04)
Yan He, Yue Zhang, Madi Sun, Chao Yang, Xiao Zheng, Chengxin Shi, Zhimin Chang, Zheng Wang, Jinying Chen, Shuchen Pei, Wen-Fei Dong, Dan Shao, Junjun She
ABSTRACT
Chlorhexidine (CHX) is a widely used antiseptic in various infection control practices. In this work, we have developed biodegradable mesoporous organosilica nanoparticles (MONs) through a one-pot synthesis by employing CHX as a bifunctional agent that not any acts as a cationic template to form the structure of mesopores but also serves as a broad-spectrum antiseptic. The resulting CHX@MONs exhibit a relatively high CHX content and glutathione (GSH)-responsive release of CHX via a matrix-degradation-controlled mechanism, leading to comparable antibacterial effects with CHX on both Escherichia coli and Staphylococcus aureus. Furthermore, the effective antibacterial concentration of CHX@MONs shows less cytotoxicity toward normal cells. Our findings will help increase the use of CHX as an antiseptic agent, especially for responsive drug release upon bacterial infection.
Sigma-Aldrich
Bis[3-(triethoxysilyl)propyl] tetrasulfide, technical, ≥90% (NMR)
Pricing and availability is not currently available.