Merck
  • Intercellular Adhesion Molecule 1 Functions as an Efferocytosis Receptor in Inflammatory Macrophages.
Intercellular Adhesion Molecule 1 Functions as an Efferocytosis Receptor in Inflammatory Macrophages.
The American journal of pathology (2020-02-09)
Hannah L Wiesolek, Triet M Bui, Joseph J Lee, Prarthana Dalal, Ariel Finkielsztein, Ayush Batra, Edward B Thorp, Ronen Sumagin
ABSTRACT
Intercellular adhesion molecule-1 (ICAM-1) is up-regulated during inflammation by several cell types. ICAM-1 is best known for its role in mediating leukocyte adhesion to endothelial cells and guiding leukocytes across the vascular wall. Recently, macrophages have been shown to express ICAM-1, however, their role in macrophage function is unclear. We found that ICAM-1 expression was induced during inflammatory macrophage polarization and high numbers of ICAM-1-expressing macrophages were noted in inflamed colon tissue in a murine colitis model and in human inflammatory bowel disease. Because tissue macrophages play a critical role in removing apoptotic/necrotic cells in inflammation and injury, a process termed efferocytosis, it was examined whether ICAM-1 contributes to this process. Genetic deletion (ICAM-1 knockout mice) or siRNA-mediated knockdown of ICAM-1 in isolated murine and human macrophages significantly impaired apoptotic cell (AC) engulfment. Impairment in the engulfment of Jurkat T cells, neutrophils, and epithelial cells was confirmed ex vivo by inflammatory macrophages and in vivo by thioglycolate-recruited peritoneal macrophages. Decreased efferocytosis was also seen in vitro and in vivo with inhibition of ICAM-1 adhesive interactions, using a function blocking anti-ICAM-1 antibody. Mechanistically, it was found that ICAM-1 actively redistributes to cluster around engulfed ACs to facilitate macrophage-AC binding. Our findings define a new role for ICAM-1 in promoting macrophage efferocytosis, a critical process in the resolution of inflammation and restoration of tissue homeostasis.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human ICAM1
Sigma-Aldrich
Triton X-100, laboratory grade