Merck
  • Protein complexes including PGRMC1 and actin-associated proteins are disrupted by AG-205.
Protein complexes including PGRMC1 and actin-associated proteins are disrupted by AG-205.
Biochemical and biophysical research communications (2020-01-26)
Sarah L Teakel, Marina Ludescher, Bashar M Thejer, Gereon Poschmann, Jade K Forwood, Hans Neubauer, Michael A Cahill
ABSTRACT
PGRMC1 is a protein from the MAPR family with a range of cellular functions. PGRMC1 has been described to play a role in fertility, neuroprotection, steroidogenesis, membrane trafficking and in cancer cell biology. PGRMC1 represents a likely key regulator of cell metabolism and proliferation, as well as a potential target for anti-cancer therapies. To further understand the functions of PGRMC1 and the mechanism of the small molecule inhibitor of PGRMC1, AG-205, proteins differentially bound to PGRMC1 were identified following AG-205 treatment of MIA PaCa-2 cells. Our results suggest that AG-205 influences PGRMC1 interactions with the actin cytoskeleton. The binding of two PGRMC1-associated proteins that support this, RACK1 and alpha-Actinin-1, was reduced following AG-205 treatment. The biology associated with PGRMC1 binding partners identified here merits further investigation.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes, ready to use solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triton X-100, laboratory grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
ColorBurst Electrophoresis Marker, mol wt 8,000-220,000 Da