- Structure and stability of phospholipid bilayers hydrated by a room-temperature ionic liquid/water solution: a neutron reflectometry study.
Structure and stability of phospholipid bilayers hydrated by a room-temperature ionic liquid/water solution: a neutron reflectometry study.
Neutron reflectometry (NR) measurements were carried out to probe the structure and stability of two model biomembranes consisting of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC) and 1,2-dimyristoyl-sn-glycero-3-phosphatidylcholine (DMPC) phospholipid bilayers hydrated by water solutions of two prototypical room-temperature ionic liquids (RTILs), namely, 1-butyl-3-methyl-imidazolium chloride ([bmim][Cl]) and choline chloride ([Chol][Cl]) at concentrations of 0.1 M and 0.5 M, respectively. The raw data were analyzed by fitting a distribution of scattering length densities arising from the different chemical species in the system. The results of this analysis show that (a) for all systems and concentrations that we considered, the thickness of the bilayers shrinks by ∼1 Å upon dissolving the ionic liquid into water and that (b) the RTIL ions enter the bilayer, finding their way to a preferred location in the lipid range that is nearly independent of the lipid and of the [bimim](+) or [Chol](+) choice. The volume fraction of RTIL sorbed in/on the bilayer, however, does depend on the lipid, but, again, is the same for [bmim][Cl] and for [Chol][Cl]. Thus, the RTIL occupies ∼5% of the bilayer volume in POPC, rising to ∼10% in DMPC. Repeating the measurements and data analysis after rinsing in pure water shows that the changes in the bilayer due to the RTIL sorption are irreversible and that a measurable amount of IL remains in the lipid fraction, that is, ∼2.5% of the bilayer volume in POPC and ∼8% in DMPC.