Merck
  • Evaluation of host-based molecular markers for the early detection of human sepsis.
Evaluation of host-based molecular markers for the early detection of human sepsis.
Journal of biotechnology (2020-02-06)
Elisabeth Ullrich, Petra Heidinger, Jung Soh, Laura Villanova, Stefan Grabuschnig, Thorsten Bachler, Elisabeth Hirschböck, Sara Sánchez-Heredero, Barry Ford, Maria Sensen, Ingund Rosales Rodriguez, Daniel Schwendenwein, Peter Neumeister, Christoph J Zurl, Robert Krause, Johannes Lorenz Khol, Christoph W Sensen
ABSTRACT
We have identified 24 molecular markers, based on circulating nucleic acids (CNA) originating from the human genome, which in combination can be used in a quantitative real-time PCR (qPCR) assay to identify the presence of human sepsis, starting two to three days before the first clinical signs develop and including patients who meet the SEPSIS-3 criteria. The accuracy was more than 87 % inside of the same patient cohort for which the markers were developed and up to 81 % in blind studies of patient cohorts which were not included in the marker development. As our markers are host-based, they can be used to capture bacterial as well as fungal sepsis, unlike the current PCR-based tests, which require species-specific primer sets for each organism causing human sepsis. Our assay directly uses an aliquot of cell-free blood as the substrate for the PCR reaction, thus allowing to obtain the diagnostic results in three to four hours after the collection of the blood samples.
Sigma-Aldrich
GenElute PCR Clean-Up Kit, sufficient for 70 purifications
Sigma-Aldrich
GenomePlex® Single Cell Whole Genome Amplification Kit, Amplify genome of a single cell