- Involvement of caspase-1 proteases in hypoxic brain injury. effects of their inhibitors in developing neurons.
Involvement of caspase-1 proteases in hypoxic brain injury. effects of their inhibitors in developing neurons.
To further explore the contribution of caspase-1/interleukin-1beta-convening enzyme in the consequences of hypoxia in developing brain neurons, its temporal expression profile was analysed by immunohistochemistry and western blotting in cultured neurons from the embryonic rat forebrain subjected to a hypoxic stress (95% N2/5% CO2 for 6 h), and proteolytic activity of caspase-1 was monitored as a function of time by measuring the degradation of a selective colorimetric substrate (N-acetyl-Tyr-Val-Ala-Asp-p-nitroanilide). In addition, the influence of pre- and posthypoxic treatments by caspase-1 inhibitors (N-acetyl-Tyr-Val-Ala-Asp-aldehyde and N-acetyl-Tyr-Val-Ala-Asp-chloromethylketone) was tested on cell outcome. Hypoxia led to delayed apoptotic neuronal death, with an elevation of the expression of both pro-caspase-1 and caspase-1 active cleavage product (ICE p20) for up to 96 h after cell reoxygenation. As reflected by cleavage of the specific substrate, caspase-1 activity progressively increased between 24 h and 96 h posthypoxia, and was blocked by inhibitors in a dose-dependent fashion. The inhibitory compounds, including when given 24 h after hypoxia, prevented neuronal death, reduced apoptosis hallmarks and also increased the number of mitotic neurons, suggesting they might promote neurogenesis. Similar observations were made when neurons were exposed to a sublethal hypoxia (i.e. 3 h). These data emphasize the participation of caspase-1 in neuronal injury consecutive to oxygen deprivation, and provide new insight into the possible cellular mechanisms by which caspase inhibitors may protect developing brain neurons.