Merck
  • A novel treatment strategy targeting polo-like kinase 1 in hematological malignancies.
A novel treatment strategy targeting polo-like kinase 1 in hematological malignancies.
Leukemia (2009-05-08)
T Ikezoe, J Yang, C Nishioka, Y Takezaki, T Tasaka, K Togitani, H P Koeffler, A Yokoyama
ABSTRACT
Polo-like kinase1 (PLK1) belongs to the family of serine/threonine kinases and plays an important role in centrosome maturation, bipolar spindle formation, and cytokinesis during mitosis. We found in this study that PLK1 was aberrantly highly expressed in a variety of human leukemia cell lines (n=20), as well as, freshly isolated leukemia cells from individuals with acute myelogenous leukemia (n=50) and acute lymphoblastic leukemia (n=15) compared with bone marrow mononuclear cells from healthy volunteers (n=13) (acute myelogenous leukemia, P=0.016; acute lymphoblastic leukemia, P=0.008), as measured by real-time RT-PCR. Downregulation of PLK1 by a small interfering RNA in NB4 acute myelogenous leukemia cells inhibited their proliferation. GW843682X is a novel selective PLK1 inhibitor. The compound-induced growth inhibition, caused accumulation of cells in the G2/M phase of the cell cycle and mediated apoptosis of human leukemia cells. Pre-treatment of cells with the caspase inhibitor Z-VAD-FMK attenuated the action of GW843682X in leukemia cells, indicating the involvement of the caspase pathway in the PLK1 inhibitor-mediated apoptosis. Furthermore, we found that the PLK1 inhibitor synergistically potentiated the growth inhibition and apoptosis of leukemia cells when combined with tubulin-depolymerizing agent vincristine. Taken together, targeting PLK1 may be a promising treatment strategy for individuals with leukemia.
Sigma-Aldrich
GW843682X, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Phosphoserine Antibody, clone 4A4 (mouse IgG1), clone 4A4, Upstate®, from mouse