Merck
  • Increased anxiety and decreased sociability induced by paternal deprivation involve the PVN-PrL OTergic pathway.
Increased anxiety and decreased sociability induced by paternal deprivation involve the PVN-PrL OTergic pathway.
eLife (2019-05-16)
Zhixiong He, Larry Young, Xin-Ming Ma, Qianqian Guo, Limin Wang, Yang Yang, Luo Luo, Wei Yuan, Laifu Li, Jing Zhang, Wenjuan Hou, Hui Qiao, Rui Jia, Fadao Tai
ABSTRACT
Early adverse experiences often have devastating consequences. However, whether preweaning paternal deprivation (PD) affects emotional and social behaviors and their underlying neural mechanisms remain unexplored. Using monogamous mandarin voles, we found that PD increased anxiety-like behavior and attenuated social preference in adulthood. PD also decreased the number of oxytocin (OT)-positive neurons projecting from the paraventricular nucleus (PVN) and reduced the levels of the medial prefrontal cortex OT receptor protein in females and of the OT receptor and V1a receptor proteins in males. Intra-prelimbic cortical OT injections reversed the PD-induced changes in anxiety-like behavior and social preferences. Optogenetic activation of the prelimbic cortex OT terminals from PVN OT neurons reversed the PD-induced changes in emotion and social preference behaviors, whereas optogenetic inhibition was anxiogenic and impaired social preference in naive voles. These findings demonstrate that PD increases anxiety-like behavior and attenuates social preferences through the involvement of PVN OT neuron projections to the prelimbic cortex.
Sigma-Aldrich
Anti-Oxytocin Antibody, clone 4G11, ascites fluid, clone 4G11, Chemicon®
Sigma-Aldrich
Anti-Vasopressin Antibody, serum, Chemicon®