Merck
  • Effects of hypoestrogenism and/or hyperaldosteronism on myocardial remodeling in female mice.
Effects of hypoestrogenism and/or hyperaldosteronism on myocardial remodeling in female mice.
Physiological reports (2018-11-16)
Patricia Rouet-Benzineb, Régine Merval, Evelyne Polidano
ABSTRACT
We investigated the potential adverse effects of hyperaldosteronism and/or hypoestrogenism on cardiac phenotype, and examined their combined effects in female mice overexpressing cardiac aldosterone synthase (AS). We focused on some signaling cascades challenging defensive responses to adapt and/or to survive in the face of double deleterious stresses, such as Ca2+ -homeostasis, pro/anti-hypertrophic, endoplasmic reticulum stress (ER stress), pro- or anti-apoptotic effectors, and MAP kinase activation, and redox signaling. These protein expressions were assessed by immunoblotting at 9 weeks after surgery. Female wild type (FWT) and FAS mice were fed with phytoestrogen-free diet; underwent ovariectomy (Ovx) or sham-operation (Sham). Ovx increased gain weight and hypertrophy index. Transthoracic echocardiograghy was performed. Both Ovx-induced heart rate decrease and fractional shortening increase were associated with collagen type III shift. Cardiac estrogen receptor (ERα, ERβ) protein expression levels were downregulated in Ovx mice. Hypoestrogenism increased plasma aldosterone and MR protein expression in FAS mice. Both aldosterone and Ovx played as mirror effects on up and downstream signaling effectors of calcium/redox homeostasis, apoptosis, such as concomitant CaMKII activation and calcineurin down-regulation, MAP kinase inhibition (ERK1/2, p38 MAPK) and Akt activation. The ratio Bcl2/Bax is in favor to promote cell survivor. Finally, myocardium had dynamically orchestrated multiple signaling cascades to restore tolerance to hostile environment thereby contributing to a better maintenance of Ca2+ /redox homeostasis. Ovx-induced collagen type III isoform shift and its upregulation may be important for the biomechanical transduction of the heart and the recovery of cardiac function in FAS mice. OVX antagonized aldosterone signaling pathways.
Sigma-Aldrich
Ponceau S, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Anti-oxidized-CaM Kinase II (Met281/282) Antibody, serum, from rabbit