Merck
  • Regulation of profibrotic responses by ADAM17 activation in high glucose requires its C-terminus and FAK.
Regulation of profibrotic responses by ADAM17 activation in high glucose requires its C-terminus and FAK.
Journal of cell science (2018-01-24)
Renzhong Li, Tony Wang, Khyati Walia, Bo Gao, Joan C Krepinsky
ABSTRACT
Glomerular matrix accumulation is the hallmark of diabetic nephropathy. The metalloprotease ADAM17 mediates high glucose (HG)-induced matrix production by kidney mesangial cells through release of ligands for the epidermal growth factor receptor. Here, we study the mechanism by which HG activates ADAM17. We find that the C-terminus is essential for ADAM17 activation and the profibrotic response to HG. In the C-terminus, Src-mediated Y702 phosphorylation and PI3K-MEK-Erk-mediated T735 phosphorylation are crucial for ADAM17 activation, both are also required for the HG-induced increase in cell surface mature ADAM17. The non-receptor tyrosine kinase FAK is a central mediator of these processes. These data not only support a crucial role for the C-terminus in ADAM17 activation and downstream profibrotic responses to HG, but also highlight FAK as a potential alternative therapeutic target for diabetic nephropathy.
Sigma-Aldrich
Anti-phospho-ADAM 17 (pThr735) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture