Merck
  • Deletion of APC7 or APC16 Allows Proliferation of Human Cells without the Spindle Assembly Checkpoint.
Deletion of APC7 or APC16 Allows Proliferation of Human Cells without the Spindle Assembly Checkpoint.
Cell reports (2018-11-30)
Thomas Wild, Magda Budzowska, Susanne Hellmuth, Susana Eibes, Gopal Karemore, Marin Barisic, Olaf Stemmann, Chunaram Choudhary
ABSTRACT
The multisubunit ubiquitin ligase APC/C (anaphase-promoting complex/cyclosome) is essential for mitosis by promoting timely degradation of cyclin B1. APC/C is tightly regulated by the spindle assembly checkpoint (SAC), which involves MPS1 and MAD2-dependent temporal inhibition of APC/C. We analyzed the contribution of the APC/C subunits APC7 and APC16 to APC/C composition and function in human cells. APC16 is required for APC7 assembly into APC/C, whereas APC16 assembles independently of APC7. APC7 and APC16 knockout cells display no major defects in mitotic progression, cyclin B1 degradation, or SAC response, but APC/C lacking these two subunits shows reduced ubiquitylation activity in vitro. Strikingly, deletion of APC7 or APC16 is sufficient to provide synthetic viability to MAD2 deletion. ΔAPC7ΔMAD2 cells display accelerated mitosis and require SAC-independent MPS1 function for genome stability. These findings reveal that the composition of APC/C critically influences the importance of the SAC in humans.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-GAPDH Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Mad2 Antibody, clone AS55-A12, clone AS55-A12, from mouse
Sigma-Aldrich
Anti-Cyclin B1 Antibody, clone GNS3 (8A5D12), clone GNS3 (8A5D12), Upstate®, from mouse
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder