- Varying label density allows artifact-free analysis of membrane-protein nanoclusters.
Varying label density allows artifact-free analysis of membrane-protein nanoclusters.
Nature methods (2016-06-14)
Florian Baumgart, Andreas M Arnold, Konrad Leskovar, Kaj Staszek, Martin Fölser, Julian Weghuber, Hannes Stockinger, Gerhard J Schütz
PMID27295310
ABSTRACT
We present a method to robustly discriminate clustered from randomly distributed molecules detected with techniques based on single-molecule localization microscopy, such as PALM and STORM. The approach is based on deliberate variation of labeling density, such as titration of fluorescent antibody, combined with quantitative cluster analysis, and it thereby circumvents the problem of cluster artifacts generated by overcounting of blinking fluorophores. The method was used to analyze nanocluster formation in resting and activated immune cells.
Sigma-Aldrich
Anti-CD3, low endotoxin antibody, Mouse monoclonal, clone OKT3, purified immunoglobulin, buffered aqueous solution