Merck
  • Propagation of trimethylated H3K27 regulated by polycomb protein EED is required for embryogenesis, hematopoietic maintenance, and tumor suppression.
Propagation of trimethylated H3K27 regulated by polycomb protein EED is required for embryogenesis, hematopoietic maintenance, and tumor suppression.
Proceedings of the National Academy of Sciences of the United States of America (2016-09-01)
Takeshi Ueda, Yuichiro Nakata, Akiko Nagamachi, Norimasa Yamasaki, Akinori Kanai, Yasuyuki Sera, Masato Sasaki, Hirotaka Matsui, Zen-Ichiro Honda, Hideaki Oda, Linda Wolff, Toshiya Inaba, Hiroaki Honda
ABSTRACT
Polycomb repressive complex 2 (PRC2) catalyzes the monomethylation, dimethylation, and trimethylation of histone H3 Lys27 (H3K27) and acts as a central epigenetic regulator that marks the repressive chromatin domain. Embryonic ectoderm development (EED), an essential component of PRC2, interacts with trimethylated H3K27 (H3K27me3) through the aromatic cage structure composed of its three aromatic amino acids, Phe97, Trp364, and Tyr365. This interaction allosterically activates the histone methyltransferase activity of PRC2 and thereby propagates repressive histone marks. In this study, we report the analysis of knock-in mice harboring the myeloid disorder-associated EED Ile363Met (I363M) mutation, analogous to the EED aromatic cage mutants. The I363M homozygotes displayed a remarkable and preferential reduction of H3K27me3 and died at midgestation. The heterozygotes increased the clonogenic capacity and bone marrow repopulating activity of hematopoietic stem/progenitor cells (HSPCs) and were susceptible to leukemia. Lgals3, a PRC2 target gene encoding a multifunctional galactose-binding lectin, was derepressed in I363M heterozygotes, which enhanced the stemness of HSPCs. Thus, our work provides in vivo evidence that the structural integrity of EED to H3K27me3 propagation is critical, especially for embryonic development and hematopoietic homeostasis, and that its perturbation increases the predisposition to hematologic malignancies.
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Histone H3 Antibody, clone 6.6.2, clone 6.6.2, Upstate®, from mouse
Sigma-Aldrich
ChIPAb+ EED - ChIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
Anti-EZH2 Antibody, clone BD43, clone BD43, from mouse
Sigma-Aldrich
Anti-EED Antibody, clone AA19, ascites fluid, from mouse