Merck
  • Allosteric Regulation of NCLX by Mitochondrial Membrane Potential Links the Metabolic State and Ca2+ Signaling in Mitochondria.
Allosteric Regulation of NCLX by Mitochondrial Membrane Potential Links the Metabolic State and Ca2+ Signaling in Mitochondria.
Cell reports (2018-12-20)
Marko Kostic, Tomer Katoshevski, Israel Sekler
ABSTRACT
Calcium is a key regulator of mitochondrial function under both normal and pathological conditions. The mechanisms linking metabolic activity to mitochondrial Ca2+ signaling remain elusive, however. Here, by monitoring mitochondrial Ca2+ transients while manipulating mitochondrial membrane potential (ΔΨm), we found that mild fluctuations in ΔΨm, which do not affect Ca2+ influx, are sufficient to strongly regulate NCLX, the major efflux pathway of Ca2+ from the mitochondria. Phosphorylation of NCLX or expression of phosphomimicking mutant (S258D) rescued NCLX activity from ΔΨm-driven allosteric inhibition. By screening ΔΨm sensitivity of NCLX mutants, we also identified amino acid residues that, through functional interaction with Ser258, control NCLX regulation. Finally, we find that glucose-driven ΔΨm changes in pancreatic β-cells control mitochondrial Ca2+ signaling primarily via NCLX regulation. Our results identify a feedback control between metabolic activity and mitochondrial Ca2+ signaling and the "safety valve" NCLX phosphorylation that can rescue Ca2+ efflux in depolarized mitochondria.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Fatty Acid-Free, Nuclease- and Protease-Free, BSA Fatty Acid-free is designed for use in serological testing, RIA, and hormone response studies. Suitable for use in Molecular Biology applications, such as Northern and Southern blots.