Merck
  • A2AR Transmembrane 2 Peptide Administration Disrupts the A2AR-A2AR Homoreceptor but Not the A2AR-D2R Heteroreceptor Complex: Lack of Actions on Rodent Cocaine Self-Administration.
A2AR Transmembrane 2 Peptide Administration Disrupts the A2AR-A2AR Homoreceptor but Not the A2AR-D2R Heteroreceptor Complex: Lack of Actions on Rodent Cocaine Self-Administration.
International journal of molecular sciences (2019-12-11)
Dasiel O Borroto-Escuela, Karolina Wydra, Wilber Romero-Fernandez, Zilong Zhou, Malgorzata Frankowska, Malgorzata Filip, Kjell Fuxe
ABSTRACT
It was previously demonstrated that rat adenosine A2AR transmembrane V peptide administration into the nucleus accumbens enhances cocaine self-administration through disruption of the A2AR-dopamine (D2R) heteroreceptor complex of this region. Unlike human A2AR transmembrane 4 (TM4) and 5 (TM5), A2AR TM2 did not interfere with the formation of the A2AR-D2R heteroreceptor complex in cellular models using BRET1 assay. A2AR TM2 was proposed to be part of the of the receptor interface of the A2AR homomer instead and was therefore tested in the current article for effects on rat cocaine self-administration using rat A2AR synthetic TM2 peptide bilaterally injected into the nucleus accumbens. The injected A2AR TM2 peptide failed to significantly counteract the inhibitory action of the A2AR agonist CGS 21680 (0.1 mg/Kg) on cocaine self-administration. In line with these results, the microinjected A2AR TM2 peptide did not reduce the number of proximity ligation assay blobs identifying A2AR-D2R heteroreceptor complexes in the nucleus accumbens. In contrast, the A2AR TM2 peptide significantly reduced the number of A2AR-A2AR homoreceptor complexes in the nucleus accumbens. As to effects on the receptor-receptor interactions in the A2AR-D2R heteroreceptor complexes, the A2AR TM2 peptide did not alter the significant increase in the D2R Ki, high values produced by the A2AR agonist CGS 21680 ex vivo in the ventral striatum. The results indicate that the accumbal A2AR-A2AR homomeric complexes are not involved in mediating the A2AR agonist-induced inhibition of cocaine self-administration.
Sigma-Aldrich
Cocaine free base
Sigma-Aldrich
Anti-Dopamine D2 receptor (DRD2) Antibody, clone 2B9, clone 3D9, from mouse
Sigma-Aldrich
Anti-Adenosine Receptor A2a Antibody, clone 7F6-G5-A2, clone 7F6-G5-A2, Upstate®, from mouse