Merck
  • Phytochemical Content, Antidiabetic, Anticholinergic, and Antioxidant Activities of Endemic Lecokia cretica Extracts.
Phytochemical Content, Antidiabetic, Anticholinergic, and Antioxidant Activities of Endemic Lecokia cretica Extracts.
Chemistry & biodiversity (2019-08-30)
Abdülmelik Aras, Ercan Bursal, Fikret Türkan, Hatice Tohma, Ömer Kılıç, İlhami Gülçin, Ekrem Köksal
ABSTRACT
The aim of this work was to investigate the enzyme inhibition, antioxidant activity, and phenolic compounds of Lecokia cretica (Lam.) DC. Acetylcholinesterase (AChE), butyrylcholinesterase (BChE), and α-glycosidase enzymes were strongly inhibited by the L. cretica extracts. IC50 values for the three enzymes were found as 3.21 mg/mL, 2.1 mg/mL, and 2.07 mg/mL, respectively. Antioxidant activities were examined in both aqueous and ethanol (EtOH) extracts using CUPRAC, FRAP, and DPPH method. Also, the phenolic compounds of the endemic plant were identified and quantified by using HPLC/MS/MS. According to the results, the extracts have remarkable antioxidant activities. The most abundant phenolic acids of L. cretica in EtOH extract were determined as quinic acid (12.76 mg/kg of crude extract), chlorogenic acid (3.39 mg/kg), and malic acid (2.38 mg/kg).
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)