Merck
  • Easy Method for the Transformation of Levulinic Acid into Gamma-Valerolactone Using a Nickel Catalyst Derived from Nanocasted Nickel Oxide.
Easy Method for the Transformation of Levulinic Acid into Gamma-Valerolactone Using a Nickel Catalyst Derived from Nanocasted Nickel Oxide.
Materials (Basel, Switzerland) (2019-09-12)
Rut Sanchis, Tomás García, Ana M Dejoz, Isabel Vázquez, Francisco J Llopis, Benjamín Solsona
ABSTRACT
Different nickel catalysts have been tested for the transformation of levulinic acid into γ-valerolactone using an easy hydrothermal method, taking advantage of the properties of the high temperature water. A metallic nickel catalyst derived from NiO synthesized by a nanocasting procedure can achieve a productivity to γ-valerolactone, which is two orders of magnitude higher than that obtained by a commercial nickel catalyst. This nanocasted metallic nickel catalyst has shown bifunctionality as it is capable of activating water as the source for hydrogen and undertaking the further hydrogenation step. In contrast with metallic nickel, nickel oxide has shown to be incapable of transforming levulinic acid into γ-valerolactone.
Sigma-Aldrich
Nickel (IV) oxide, technical, oxidizing agent, ~30% active peroxide basis