Merck
  • Serum electrolytes can promote hydroxyl radical-initiated biomolecular damage from inflammation.
Serum electrolytes can promote hydroxyl radical-initiated biomolecular damage from inflammation.
Free radical biology & medicine (2019-07-28)
Yukako Komaki, Adam M-A Simpson, Jong Kwon Choe, Margaux M Pinney, Daniel Herschlag, Yi-Hsueh Chuang, William A Mitch
ABSTRACT
Chronic inflammatory disorders are associated with biomolecular damage attributed partly to reactions with Reactive Oxygen Species (ROS), particularly hydroxyl radicals (•OH). However, the impacts of serum electrolytes on ROS-associated damage has received little attention. We demonstrate that the conversion of •OH to carbonate and halogen radicals via reactions with serum-relevant carbonate and halide concentrations fundamentally alters the targeting of amino acids and loss of enzymatic activity in catalase, albumin and carbonic anhydrase, three important blood proteins. Chemical kinetic modeling indicated that carbonate and halogen radical concentrations should exceed •OH concentrations by 6 and 2 orders of magnitude, respectively. Steady-state γ-radiolysis experiments demonstrated that serum-level carbonates and halides increased tyrosine, tryptophan and enzymatic activity losses in catalase up to 6-fold. These outcomes were specific to carbonates and halides, not general ionic strength effects. Serum carbonates and halides increased the degradation of tyrosines and methionines in albumin, and increased the degradation of histidines while decreasing enzymatic activity loss in carbonic anhydrase. Serum electrolytes increased the degradation of tyrosines, tryptophans and enzymatic activity in the model enzyme, ketosteroid isomerase, predominantly due to carbonate radical reactions. Treatment of a mutant ketosteroid isomerase indicated that preferential targeting of the active site tyrosine accounted for half of the total tyrosine loss. The results suggest that carbonate and halogen radicals may be more significant than •OH as drivers for protein degradation in serum. Accounting for the selective targeting of biomolecules by these daughter radicals is important for developing a mechanistic understanding of the consequences of oxidative stress.
Sigma-Aldrich
N-Acetyl-L-leucine, ReagentPlus®, 99%
Sigma-Aldrich
Nα-Acetyl-L-lysine
Sigma-Aldrich
Nα-Acetyl-L-arginine
Sigma-Aldrich
Nα-Acetyl-L-glutamine