Merck
Programmed cell death in soybean seed coats.
Plant science : an international journal of experimental plant biology (2019-09-16)
Gustavo Lemos Rocha, Saulo Pireda, Jucélia da Silva Araújo, Antônia Elenir Amâncio Oliveira, Olga Lima Tavares Machado, Maura da Cunha, Clicia Grativol, Kátia Valevski Sales Fernandes
ABSTRACT
Seed coat is the tissue which establishes an interface between the seed inner tissues and external environment. Our group has shown that cowpea seed coat undergoes coordinated events of programmed cell death (PCD) during development. In relation to germinating seed coats, little is known on PCD events. The goal here was to investigate the biochemical aspects of germinating soybean seed coat, focusing on proteolytic activities related to PCD. In gel and in solution activity profiles of quiescent and germinating seed coat extracts revealed a complex pattern of caspase- and metacaspase-like cysteine protease activities. Trypsin inhibitor and reserve proteins were revealed as potential substrates for these proteases. A pancaspase inhibitor (z-VAD-CHO) affected the radicle length of seeds germinated under its presence. Ultrastructural analysis showed the absence of cell organelles in all seed coat layers after imbibition, while oligonucleosome fragments peaked at 72 h after imbibition (HAI). Altogether, the data suggest the presence of biochemical PCD hallmarks in germinating soybean seed coat and point to the involvement of the detected protease activities in processes such as reserve protein mobilization and weakening of seed coat.
Sigma-Aldrich
Toluidine Blue, for microscopy (Hist., Vit.), 8.74% (ZN (THEORY))
Sigma-Aldrich
N-Acetyl-Val-Glu-Ile-Asp-al, ~99% (HPLC), powder
Sigma-Aldrich
N-Acetyl-Tyr-Val-Ala-Asp-7-amido-4-trifluoromethylcoumarin, ≥95% (HPLC), powder
Sigma-Aldrich
Human SPINT1 / Kunitz-type Protease Inhibitor 1 ELISA Kit