Merck
  • The JAK2 inhibitors CEP-33779 and NVP-BSK805 have high P-gp inhibitory activity and sensitize drug-resistant cancer cells to vincristine.
The JAK2 inhibitors CEP-33779 and NVP-BSK805 have high P-gp inhibitory activity and sensitize drug-resistant cancer cells to vincristine.
Biochemical and biophysical research communications (2017-07-04)
Ji Hyun Cheon, Kyeong Seok Kim, Dharmendra Kumar Yadav, Mihyun Kim, Hyung Sik Kim, Sungpil Yoon
ABSTRACT
P-glycoprotein (P-gp) is overexpressed in cancer cells in order to pump out chemotherapeutic drugs, and is one of the major mechanisms responsible for multidrug resistance (MDR). It is important to identify P-gp inhibitors with low toxicity to normal cells in order to increase the efficacy of anti-cancer drugs. Previously, a JAK2 inhibitor CEP-33779 demonstrated inhibitory actions against P-gp and an ability to sensitize drug-resistant cancer cells to treatment. In the present study, we tested another JAK2 inhibitor NVP-BSK805 for P-gp inhibitory activity. In molecular docking simulation modeling, NVP-BSK805 showed higher binding affinity docking scores against a P-gp member (ABCB1) than CEP-33779 did. Furthermore, we found that lower doses of NVP-BSK805 are required to inhibit P-gp in comparison with that of CEP-33779 or verapamil (an established P-gp inhibitor) in KBV20C cells, suggesting that NVP-BSK805 has higher specificity. NVP-BSK805, CEP-33779, and verapamil demonstrated similar abilities to sensitize KBV20C cells to vincristine (VIC) treatment. Our results suggested that the JAK2 inhibitors were able to inhibit P-gp pump-action via a direct binding mechanism, similar to verapamil. However, JAK2 inhibitor-induced sensitization was not observed in VIC-treated sensitive KB parent cells, suggesting that these effects are specific to resistant cancer cells. FACS, western-blot, and annexin V analyses were used to further investigate the mechanism of action of JAK2 inhibitors in VIC-treated KBV20C cells. Both CEP-33779 and NVP-BSK805 induced the sensitization of KBV20C cells to VIC treatment via the same mechanisms; they each caused a reduction in cell viability, increased G2 arrest, and upregulated expression of the DNA damaging protein pH2AX when used as co-treatments with VIC. These findings indicate that inhibition of JAK2 may be a promising target in the treatment of cancers that are resistant to anti-mitotic drugs.
Sigma-Aldrich
NVP-BSK805 Trihydrochloride, ≥98% (HPLC)