Merck
  • Photoreduction of triplet thioxanthone derivative by azolium tetraphenylborate: a way to photogenerate N-heterocyclic carbenes.
Photoreduction of triplet thioxanthone derivative by azolium tetraphenylborate: a way to photogenerate N-heterocyclic carbenes.
Physical chemistry chemical physics : PCCP (2019-07-30)
Thi Kim Hoang Trinh, Fabrice Morlet-Savary, Julien Pinaud, Patrick Lacroix-Desmazes, Corine Reibel, Cécile Joyeux, Didier Le Nouen, Rémi Métivier, Arnaud Brosseau, Valérie Héroguez, Abraham Chemtob
ABSTRACT
Although N-heterocyclic carbenes (NHCs) have brought profound changes in catalytic organic synthesis, their generation generally requires an inert atmosphere and harsh conditions. To overcome these limitations, an air-stable NHC photogenerator has been developed involving two mild components: 1,3-bis(mesityl)imidazolium tetraphenylborate (IMesH+BPh4-) and electronically excited isopropylthioxanthone (ITX). In this study, the photochemical mechanism is investigated via the accurate identification of the transient species and photoproducts. Electron transfer reaction between the excited triplet state of ITX and BPh4- is demonstrated as being the primary photochemical step. Nanosecond laser spectroscopy shows an efficient quenching and the formation of the expected ITX radical anion. The oxidized borane species is not observed, suggesting that this short-lived species could dissociate very rapidly to give the phenyl radical - successfully identified using electron paramagnetic resonance - and triphenylborane. As regards the final photoproducts, 1H and 13C NMR spectroscopies support the formation of the targeted NHC, 1,3-bis(2,4,6-trimethylphenyl)imidazol-2-ylidene (IMes), suggesting the occurrence of a subsequent proton transfer reaction between ITX radical anion and imidazolium cation (IMesH+). Gas chromatography-mass spectrometry reveals three other products: biphenyl, isopropylthioxanthene and ITX. Their formation can be reconciled with a 2-step mechanism of photoinduced electron/proton transfer reactions. 11B NMR spectroscopy demonstrates that the main organoboron photoproduct is diphenylborinic acid formed by oxidation of BPh3. Due to its Lewis acidity, Ph2BOH can react with IMes to yield an NHC-boron adduct.