Merck
  • A RASSF1A-HIF1α loop drives Warburg effect in cancer and pulmonary hypertension.
A RASSF1A-HIF1α loop drives Warburg effect in cancer and pulmonary hypertension.
Nature communications (2019-05-16)
Swati Dabral, Christian Muecke, Chanil Valasarajan, Mario Schmoranzer, Astrid Wietelmann, Gregg L Semenza, Michael Meister, Thomas Muley, Tamina Seeger-Nukpezah, Christos Samakovlis, Norbert Weissmann, Friedrich Grimminger, Werner Seeger, Rajkumar Savai, Soni S Pullamsetti
ABSTRACT
Hypoxia signaling plays a major role in non-malignant and malignant hyperproliferative diseases. Pulmonary hypertension (PH), a hypoxia-driven vascular disease, is characterized by a glycolytic switch similar to the Warburg effect in cancer. Ras association domain family 1A (RASSF1A) is a scaffold protein that acts as a tumour suppressor. Here we show that hypoxia promotes stabilization of RASSF1A through NOX-1- and protein kinase C- dependent phosphorylation. In parallel, hypoxia inducible factor-1 α (HIF-1α) activates RASSF1A transcription via HIF-binding sites in the RASSF1A promoter region. Vice versa, RASSF1A binds to HIF-1α, blocks its prolyl-hydroxylation and proteasomal degradation, and thus enhances the activation of the glycolytic switch. We find that this mechanism operates in experimental hypoxia-induced PH, which is blocked in RASSF1A knockout mice, in human primary PH vascular cells, and in a subset of human lung cancer cells. We conclude that RASSF1A-HIF-1α forms a feedforward loop driving hypoxia signaling in PH and cancer.
Sigma-Aldrich
Monoclonal Anti-Collagen, Type I antibody produced in mouse, clone COL-1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid