Merck
  • Self-Assembly of Stable Nanoscale Platelets from Designed Elastin-like Peptide-Collagen-like Peptide Bioconjugates.
Self-Assembly of Stable Nanoscale Platelets from Designed Elastin-like Peptide-Collagen-like Peptide Bioconjugates.
Biomacromolecules (2019-02-23)
Jingya Qin, Tianzhi Luo, Kristi L Kiick
ABSTRACT
The self-assembly of nanostructures from elastin-like (poly)peptide (ELP) containing block copolymers has been a subject of intense investigation over decades. However, short synthetic ELPs have rarely been used due to their high inverse transition temperature; the use of short ELPs has largely been limited to polymer conjugates. Motivated by our previous work which successfully overcame this barrier by simply conjugating short ELPs with a triple-helix-forming collagen-like peptide, in this study, we further extend the ELP library to a series of ELPs equipped with aromatic residues and having sequences as short as four pentapeptide motifs. The resulting elastin-like peptide-collagen-like peptide (ELP-CLP) bioconjugates unexpectedly self-assembled into nanosized platelets likely by forming a bilayer structure. Given the previously demonstrated ability of many other CLP conjugates to target collagens and the potential for encapsulation of hydrophobic drugs in collapsed ELPs, these ELP-CLP nanoplatelets may offer similar opportunities for targeted delivery in biomedical and other arenas.