Merck
  • Micellar system in copper-catalysed hydroxylation of arylboronic acids: facile access to phenols.
Micellar system in copper-catalysed hydroxylation of arylboronic acids: facile access to phenols.
Chemical communications (Cambridge, England) (2011-10-01)
Kiyofumi Inamoto, Kanako Nozawa, Misato Yonemoto, Yoshinori Kondo
ABSTRACT
Copper-catalysed oxidative hydroxylation of arylboronic acids was accomplished in water containing an amphiphilic surfactant, providing facile access to phenol derivatives.
Sigma-Aldrich
4-Iodophenylboronic acid, ≥95.0%