Merck
  • Association between HLA-DRB1* polymorphisms and hepatitis B infection in a brazilian population.
Association between HLA-DRB1* polymorphisms and hepatitis B infection in a brazilian population.
Revista da Associacao Medica Brasileira (1992) (2012-10-24)
Bruno de Melo Corrêa, Edmundo Pessoa de Almeida Lopes, Maria de Fátima Pessoa Militão de Albuquerque, Lúcia Dourado
ABSTRACT
The aim of the present study was to determine the genotype association for alleles of class II human leukocyte antigens (HLA) in the DRB1* locus among blood donors at the Fundação Hemope (Brazil) infected by or immunized for the hepatitis B virus (HBV). A case-control study was performed, comprising a group of individuals infected by HBV and a control group of immunized individuals at a proportion of 1:4. Blood samples were taken for the HLA typing of the DRB1* locus. Univariate and multivariate analyses were performed for the assessment of associations between the categorical variables using the chi-squared test and Fisher's exact test. A total of 320 blood donors were analyzed (241 males [75%] and 79 females [25%] with a mean age of 39 years). The case group consisted of 64 HBV-infected donors and the control group was composed of 256 HBV-immunized donors. The multivariate analysis stratified by gender revealed that the DRB1*09 allele was associated with infected male donors (p = 0.016) and the DRB1*08 allele was associated with infected donors aged 39 years or younger (p = 0.031). The results of the present study reveal that younger blood donors and male blood donors who respectively exhibit the DRB1*08 and DRB1*09 alleles are more susceptible to intensification of HBV infection.